Propylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

P2212-1G P2212-5G
TCIAP2212-1GEA 70 EUR
TCIAP2212-1G TCIAP2212-5G
Propylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
Propylamine hydroiodide
Formula: C₃H₉N
MW: 187.02 g/mol
Núm. CAS: 14488-45-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8abf4c0516148c9f48510d34ed98fc1fd16e55b&vid=e8abf4c0516148c9f48510d34ed98fc1fd16e55b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR