Ftalocianina for organic electronics, purified by sublimation

Proveedor: TCI

P2734-100MG P2734-500MG
TCIAP2734-100MGEA 67 EUR
TCIAP2734-100MG TCIAP2734-500MG
Ftalocianina for organic electronics, purified by sublimation
Ftalocianina
Formula: C₃₂H₁₈N₈
Número MDL: MFCD00005085
Núm. CAS: 574-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR