Rhodamine 800 ≥98.0% (by HPLC)

Proveedor: TCI
Atención

R0190-200MG
TCIAR0190-200MGEA 295 EUR
TCIAR0190-200MG
Rhodamine 800 ≥98.0% (by HPLC)
Rhodamine 800
Formula: C₂₆H₂₆ClN₃O₅
MW: 495.96 g/mol
Número MDL: MFCD00467020
Núm. CAS: 137993-41-0
UN: 1479
ADR: 5.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aa6a1c4c43e6b390ee89cb74ee397d9ceccf972&vid=5aa6a1c4c43e6b390ee89cb74ee397d9ceccf972&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Green to Dark green to Dark blue powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Lambda max. 677.0 to 681.0 nm (MeOH)
Absorbance (E1%1cm) min. 2000 (MeOH, 677 to 681 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR