Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI

Sinónimos: Methyl DL-serinate hydrochloride, H-DL-Ser-OMe·HCl, 3-hydroxy-1-methoxy-1-oxopropan-2-aminium chloride, methyl 2-amino-3-hydroxypropanoate, DL-Serine Methyl Ester Hydrochloride, H-DL-Ser-OMe.HCl

S0576-25G S0576-5G
TCIAS0576-25GEA 48 EUR
TCIAS0576-25G TCIAS0576-5G
Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Methyl-2-amino-3-hydroxypropanoate hydrochloride
Formula: C₄H₁₀ClNO₃
MW: 155.578 g/mol
Número MDL: MFCD00012593
Núm. CAS: 5619-04-5
EINECS: 227-047-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR