Tetraphenylarsonium chloride hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

T0162-1G
TCIAT0162-1GEA 97 EUR
TCIAT0162-1G
Tetraphenylarsonium chloride hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)
Tetraphenylarsonium chloride hydrochloride hydrate
Formula: (C₆H₅)₄AsCl·HCl·xH₂O
Número MDL: MFCD00012628
Núm. CAS: 123334-18-9
UN: 3465
ADR: 6.1,III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR