D-Treonina ≥98.0% (by titrimetric analysis)

Proveedor: TCI

T0228-25G T0228-500G T0228-100G
TCIAT0228-25GEA 42 EUR
TCIAT0228-25G TCIAT0228-500G TCIAT0228-100G
D-Treonina ≥98.0% (by titrimetric analysis)
D-Treonina
Formula: C₄H₉NO₃
Número MDL: MFCD00064269
Núm. CAS: 632-20-2
EINECS: 211-171-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=268ec1309e7ee25882df3f63b6af9d7441c2025f&vid=268ec1309e7ee25882df3f63b6af9d7441c2025f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +27.0 to +29.0 deg (C=3, H2O)
Solubility in Water almost transparency

ESPECIFICACIONES

Environmentally Preferable Safer Human and Environmental Health

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR