(Trimethylsilyl)diazomethane ∼10% (w/w) ∼0.6 mol/L in hexane

Proveedor: TCI
Peligro

T1146-10ML T1146-100ML T1146-25ML
TCIAT1146-10MLEA 76 EUR
TCIAT1146-10ML TCIAT1146-100ML TCIAT1146-25ML
(Trimethylsilyl)diazomethane ∼10% (w/w) ∼0.6 mol/L in hexane
(Trimethylsilyl)diazomethane
Formula: C₄H₁₀N₂Si
Temperatura de almacenaje: Nevera
Número MDL: MFCD00053946
Núm. CAS: 18107-18-1
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cab81309b6472e72fcc16415885d59863a05fff8&vid=4ed6393819d3a492519bb168c9f6d46577eeca18&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow clear liquid
Concentration (Neutralization titration) 10.0 to 11.0 w/w%
High boiling impurities to pass test

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR