Tetrabutylammonium dichloroaurate

Proveedor: TCI
Atención

T1273-100MG
TCIAT1273-100MGEA 125 EUR
TCIAT1273-100MG
Tetrabutylammonium dichloroaurate
Tetrabutylammonium dichloroaurate
Formula: C₁₆H₃₆AuCl₂N
Número MDL: MFCD00059125
Núm. CAS: 50480-99-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68143f8a84dc5f3337c835673e0c38ed64af8a48&vid=68143f8a84dc5f3337c835673e0c38ed64af8a48&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Melting point 80.0 to 84.0 deg-C
Ignition residue (as Au) 37.0 to 39.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR