Trifluorometanosulfonato de litio ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

T1548-25G
TCIAT1548-25GEA 77 EUR
TCIAT1548-25G
Trifluorometanosulfonato de litio ≥98.0% (by titrimetric analysis)
Trifluorometanosulfonato de litio
Formula: CF₃LiO₃S
MW: 156.01 g/mol
Número MDL: MFCD00013227
Núm. CAS: 33454-82-9
EINECS: 251-528-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07f9ef919234dad22b13327f838a819abed58338&vid=07f9ef919234dad22b13327f838a819abed58338&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR