1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: [1,2,4]Triazolo[4,3-a]pyridin-3(2H)-one, 3-Hydroxy-1,2,4-triazolo[4,3-a]pyridine

T1667-25G
TCIAT1667-25GEA 43 EUR
TCIAT1667-25G
1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one ≥98.0% (by HPLC, titration analysis)
1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one
Formula: C₆H₅N₃O
MW: 135,13 g/mol
Número MDL: MFCD00022632
Núm. CAS: 6969-71-7
EINECS: 230-191-8

Order Now

Resultados test especificaciones

Appearance White to Green to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 230.0 to 234.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR