N,N,N',N'-Tetrakis(p-tolyl)benzidine ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

T2269-5G
TCIAT2269-5GEA 99 EUR
TCIAT2269-5G
N,N,N',N'-Tetrakis(p-tolyl)benzidine ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetrakis(p-tolyl)benzidine
Formula: C₄₀H₃₆N₂
Número MDL: MFCD00799300
Núm. CAS: 76185-65-4

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 216.0 to 220.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR