1-(4-(Trifluorometil)fenil)piperazina ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

T2741-25G T2741-5G
TCIAT2741-25GEA 247 EUR
TCIAT2741-25G TCIAT2741-5G
1-(4-(Trifluorometil)fenil)piperazina ≥98.0% (by GC, titration analysis)
1-(4-(Trifluorometil)fenil)piperazina
Formula: C₁₁H₁₃F₃N₂
Número MDL: MFCD00040765
Núm. CAS: 30459-17-7
EINECS: 250-210-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9685a368ae6d085e3eb41813e3cebce68360a67f&vid=9685a368ae6d085e3eb41813e3cebce68360a67f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR