Tianeptine sodium salt hydrate ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

T3131-200MG T3131-1G
TCIAT3131-200MGEA 71 EUR
TCIAT3131-200MG TCIAT3131-1G
Tianeptine sodium salt hydrate ≥98.0% (by HPLC, total nitrogen)
Tianeptine sodium salt hydrate
Formula: C₂₁H₂₄ClN₂NaO₄S·xH₂O
Número MDL: MFCD00879851
Núm. CAS: 30123-17-2
EINECS: 250-059-3
Índice Merck: 14,09420
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec5c1692eb584f7f9dfe0bc2fb17de2c86916f01&vid=b9cd0e26ce542496828e343bd37ebd312c4a1fb0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Melting point 183.0 to 187.0 deg-C
Water 3.0 to 7.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR