2-Naphthyl boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

T3298-1G T3298-5G
TCIAT3298-1GEA 24 EUR
TCIAT3298-1G TCIAT3298-5G
2-Naphthyl boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Naphthyl boronic acid pinacol ester
Formula: C₁₆H₁₉BO₂
Número MDL: MFCD05663868
Núm. CAS: 256652-04-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0983345fe2de2aacfc9f872954e56f2782aa088e&vid=0983345fe2de2aacfc9f872954e56f2782aa088e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR