3-(Trifluoromethoxy)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

T3377-1G T3377-5G
TCIAT3377-1GEA 40 EUR
TCIAT3377-1G TCIAT3377-5G
3-(Trifluoromethoxy)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-(Trifluoromethoxy)phenylboronic acid pinacol ester
Formula: C₁₃H₁₆BF₃O₃
Número MDL: MFCD05863919
Núm. CAS: 262376-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c50aeac4ba41769109c007a717093d22fb9b01df&vid=c50aeac4ba41769109c007a717093d22fb9b01df&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR