Cole Parmer® Dri-Block® BH-250 Heaters
Proveedor: Cole-Parmer
36620-00 36620-10 36620-04 36620-02 36620-06 36620-08 36620-12 36620-14 36620-16
TECN36620-00EA 973 EUR
TECN36620-00 TECN36620-10 TECN36620-04 TECN36620-02 TECN36620-06 TECN36620-08 TECN36620-12 TECN36620-14 TECN36620-16
Cole Parmer® Dri-Block® BH-250 Heaters
Termobloques Calentadores de bloque digitales
Choose a heater, then blocks to suit your application needs!
  • Precision-engineered heaters with excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Take up less storage space-lightweight and flat-top surface allow units to be stacked when not in use
Use these heaters as a dry, constant temperature source for various lab and research applications. All models have independent indicator lights to show main power, heater action, and over-temperature cut out.
The 'number of blocksMore Product Information
Order Now
ESPECIFICACIONES
Modelo 36620-00 36620-10 36620-04 36620-02 36620-06 36620-08 36620-12 36620-14 36620-16
Environmentally Preferable
Intervalo de temperaturas 100 °C 200 °C 100 °C 100 °C 100 °C 200 °C 100 °C 200 °C 200 °C
Uniformidad de la temperatura 0,2 °C within the block at 37 °C; 0,5 °C between block at 37 °C 0,2 °C within the block at 37 °C; 0,5 °C between block at 37 °C 0,2 °C within the block at 37 °C; 0,5 °C between block at 37 °C 0,2 °C within the block at 37 °C; 0,5 °C between block at 37 °C 0,2 °C within the block at 37 °C; 0,5 °C between block at 37 °C 0,2 °C within the block at 37 °C; 0,5 °C between block at 37 °C 0,2 °C at 40 °C within identical blocks 0,2 °C at 40 °C within identical blocks 0,2 °C within the block at 37 °C; 0,5 °C between block at 37 °C
Alimentación 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz