Hygienox Electropolished Seating Benches
Proveedor: TEKNOMEK
CRSEHYEPSB1035 CRSEHYEPSB1235 CRSEHYEPSB1535 CRSEHYEPSB1835 CRSEHYEPSB2035 CRSEHYEPSB2535
TEKMCRSEHYEPSB1035EA 0 EUR
TEKMCRSEHYEPSB1035 TEKMCRSEHYEPSB1235 TEKMCRSEHYEPSB1535 TEKMCRSEHYEPSB1835 TEKMCRSEHYEPSB2035 TEKMCRSEHYEPSB2535
Hygienox Electropolished Seating Benches
Mobiliario Mesas
Hygienox electropolished 304L-grade stainless steel seating benches are made specifically for cleanroom environments, giving extreme protection against pitting and crevice corrosion caused by harsh clean down routines.
  • Optimum sitting height
  • Easy to swivel over
  • Four stainless steel adjustable feet with non-marking rubber base
  • Use a joining bracket to create a larger bench
Providing protection from corrosive chemicals, such as chlorine and hydrogen peroxide, the hygienox seating benches are also easier to clean due to the smoother surface of theMore Product Information