Tables/workbenches with multi-bar under shelf for optimum airflow
Proveedor: TEKNOMEK
MTSS0600X600/S MTSS0900X600/S MTSS1200X600/S MTSS1200X750/S MTSS1500X600/S MTSS1500X750/S MTSS1500X900/S MTSS1800X600/S MTSS1800X750/S MTSS1800X900/S MTSSB0600X600S MTSSB0900X600S MTSSB1200X600S MTSSB1200X750S MTSSB1500X600S MTSSB1500X750S MTSSB1500X900S MTSSB1800X600S MTSSB1800X750S MTSSB1800X900S TSS0600X600 TSS0900X600 TSS1200X600 TSS1200X750 TSS1500X600 TSS1500X750 TSS1500X900 TSS1800X600/4 TSS1800X750/4 TSS1800X900/4 TSSB0600X600 TSSB0900X600 TSSB1200X600 TSSB1200X750 TSSB1500X600 TSSB1500X750 TSSB1500X900 TSSB1800X600/4 TSSB1800X750/4 TSSB1800X900/4
TEKMMTSS0600X600/SEA 0 EUR
TEKMMTSS0600X600/S TEKMMTSS0900X600/S TEKMMTSS1200X600/S TEKMMTSS1200X750/S TEKMMTSS1500X600/S TEKMMTSS1500X750/S TEKMMTSS1500X900/S TEKMMTSS1800X600/S TEKMMTSS1800X750/S TEKMMTSS1800X900/S TEKMMTSSB0600X600S TEKMMTSSB0900X600S TEKMMTSSB1200X600S TEKMMTSSB1200X750S TEKMMTSSB1500X600S TEKMMTSSB1500X750S TEKMMTSSB1500X900S TEKMMTSSB1800X600S TEKMMTSSB1800X750S TEKMMTSSB1800X900S TEKMTSS0600X600 TEKMTSS0900X600 TEKMTSS1200X600 TEKMTSS1200X750 TEKMTSS1500X600 TEKMTSS1500X750 TEKMTSS1500X900 TEKMTSS1800X600/4 TEKMTSS1800X750/4 TEKMTSS1800X900/4 TEKMTSSB0600X600 TEKMTSSB0900X600 TEKMTSSB1200X600 TEKMTSSB1200X750 TEKMTSSB1500X600 TEKMTSSB1500X750 TEKMTSSB1500X900 TEKMTSSB1800X600/4 TEKMTSSB1800X750/4 TEKMTSSB1800X900/4
Tables/workbenches with multi-bar under shelf for optimum airflow
Mobiliario Mesas
This stainless steel workbench features a multi-bar under shelf which helps maintain cleanroom airflow and is perfect for storing items out of the way, but close to hand for when they are needed.
  • Fully welded 304-grade stainless steel, brushed dull polish finish
  • Supplied fully welded, no assembly
  • Multi-bar under shelf for optimum airflow
  • Choose from static or mobile with BZP or stainless steel castors
  • Added strength
304 grade stainless steel, sturdy 40 mm box section frame, designed with hygiene in mind.
Load capacity: 225 kg (for models withMore Product Information