Label holders, clip on, for freezer racks
Proveedor: TENAK
TE76100 TE76110 TE76120
479-1324EA 14.2 EUR
479-1324 479-1325 TENATE76120
Label holders, clip on, for freezer racks
Portaetiquetas
These clip on label holders are ideal for easy organizing of freezers.
  • With handle lifter to grab the rack handle
  • Easy to install, clip onto the rack and the barb secures it
The holder can be used for racks for both upright freezers and chest freezers. It is universal and can be used for most commonly sold racks of both stainless steel and aluminum with a material thickness of up to 1,5 mm.