Infrared thermometer, 830
Proveedor: Testo
0560.8316 0563.8315
TEST0560.8316EA 153 EUR
TEST0560.8316 TEST0563.8315
Infrared thermometer, 830
Termómetros Termómetros digitales
The testo 831 is so fast that measurements on palettes or refrigerated shelves can be carried out in a few seconds. Small objects such as yoghurt pots can be measured at a distance. A two-point laser indicates the exact measurement spot.
  • Adjustable emissivity of 0,2 to 1,0
  • 30:1 optics (measurement spot Ø is 36 mm at a distance of one metre)
  • Alarm limit values can be set and are optically and audibly indicated
Certificaciones: HACCP conform
Información de suministro: Supplied with a belt holder, a nine-volt battery for 15 hours of use, and a factoryMore Product Information