Thermal imagers, testo 883
Proveedor: Testo
0560.8830 0563.8830 0560.8831 0563.8831
TEST0560.8830EA 4133 EUR
TEST0560.8830 TEST0563.8830 TEST0560.8831 TEST0563.8831
Thermal imagers, testo 883
Cámaras
The testo 883 thermal imager provides outstanding image quality, automatic allocation/archiving of thermal images to the measurement object and professional image analysis and reporting.
  • Useful features: testo SiteRecognition intelligent image management and testo ScaleAssist automatic contrast adjustment
  • Extensive analysis and documentation: With the intuitive professional software testo IRSoft
  • Full control: Manual focus and interchangeable lenses
  • Interchangeable telephoto lens for distant objects and outdoor recording
  • Manual focus for sharp thermal