TexVantage™ Dry Polyester Cleanroom Wipers, Non-Sterile
Proveedor: Texwipe
TX8949
TEXWTX8949EA 0 EUR
TEXWTX8949
TexVantage™ Dry Polyester Cleanroom Wipers, Non-Sterile
Toallitas Toallitas para salas limpias Seco
TexVantage™ Polyester cleanroom wipers are constructed from 100% polyester with a sealed-edge, creating a wiper ideal for spill control, cleaning and solution application. They are designed for use on abrasive surfaces. This wiper will not easily snag or abrade releasing particles and fibers into the process or environment.
  • Autoclave safe
  • 100% Polyester with a Sealed-Edge
  • Excellent chemical resistance for compatibility with a variety of solutions
  • Meets USP <797> and USP <800> wiper requirements
  • Lining trays for holding, protecting, drying and