TSKgel® gel filtration columns, PW-type
Proveedor: TOSOH Bioscience
0005105 0005106 0005107 0005108 0005109 0005761 0005762 0005763 0005764 0005765 0006758 0006762 0006763 0008020 0008021 0008022 0008023 0008024 0008025 0008026 0008027 0008028 0008029 0008030 0008031 0008032 0008033 0008034 0021873 0021874 0021875 0021876 0020024 0022790 0022789 0022791 0022792 0022793 0022794 0022795 0022796
TOYO0005105EA 3540 EUR
TOYO0005105 TOYO0005106 TOYO0005107 554-1604 TOYO0005109 554-1607 554-1608 554-1609 554-1610 554-1653 554-1654 554-1611 554-1655 554-1391 554-1406 554-1407 554-1408 554-1409 554-1410 554-1616 TOYO0008027 TOYO0008028 TOYO0008029 TOYO0008030 554-1617 TOYO0008032 554-1411 554-1618 TOYO0021873 TOYO0021874 TOYO0021875 TOYO0021876 TOYO0020024 TOYO0022790 TOYO0022789 TOYO0022791 TOYO0022792 TOYO0022793 TOYO0022794 TOYO0022795 TOYO0022796
TSKgel® gel filtration columns, PW-type
Columnas de cromatografía
These columns are designed for GFC of water soluble organic polymers, polysaccharides, oligosaccharides, DNA and RNA. They are based on a hydrophilic polymethacrylate matrix, which is stable from pH 2 to 12 and in aqueous eluents with up to 50% polar organic solvent.
  • TSKgel® PWXL range is preferred for analytical purposes, due to the increased efficiency and resolution
  • The 600 mm length TSKgel® PW columns are recommended for their increased loading capacity for preparative work
  • The mixed bed TSKgel® GMPW and GMPWXL columns are useful for samples with a