TSKgel® reversed phase columns
Proveedor: TOSOH Bioscience
0007125 0007190 0007637 0008043 0013351 0014005 0017201 0017202 0017500 0018151 0018154 0018197 0018207 0019541 0020014 0018276 0019542 0020013 0018275 0018150 0018277 0018278 0022827 0022828 0022829 0022830 0022831 0022832 0022833 0022834
554-1613EA 726 EUR
554-1613 554-1614 554-1615 554-1619 554-0909 554-0910 554-1626 554-1627 554-0913 554-1642 554-0915 554-0916 554-1644 554-0923 554-0926 554-0918 554-0924 554-0925 554-0917 554-1641 554-1647 554-1648 TOYO0022827 TOYO0022828 TOYO0022829 TOYO0022830 TOYO0022831 TOYO0022832 TOYO0022833 TOYO0022834
TSKgel® reversed phase columns
Columnas de cromatografía
These silica- and polymer-based columns are ideal for reversed-phase applications such as pharmaceutical drugs, forensic compounds, derivatised amino acids, carbohydrates, steroids, lipids and fatty acids. Depending on their molecular weight, peptides may be analysed with the 80 to 140 Å silica-based columns, while proteins require a larger pore size column (200 to 300 Å).
  • Silica and polymer-based materials
  • Super series ideal for LCMS
  • High recovery for biomolecules
TSKgel® Protein C4-300:
TSKgel® Protein C4-300 consists of a polymerically bonded butyl (More Product Information