SkillPak™ BIO Pre-Packed Columns
Proveedor: TOSOH Bioscience
0045346 0045347 0045348 0045349 0045350 0045351 0045352 0045353 0045354 0045355 0045356 0045357 0045358 0045359 0045360 0045361 0045362 0045363 0045364 0045365 0045366 0045367 0045368 0045369 0045370 0045371 0045372 0045373 0045374 0045375 0045376 0045377 0045378 0045379 0045380 0045381 0045382 0045383 0045384 0045385 0045386 0045387 0045388 0045389 0045390 0045392 0045393 0045394 0045395 0045396 0045397
TOYO0045346EA 2820 EUR
TOYO0045346 TOYO0045347 TOYO0045348 TOYO0045349 TOYO0045350 TOYO0045351 TOYO0045352 TOYO0045353 TOYO0045354 TOYO0045355 TOYO0045356 TOYO0045357 TOYO0045358 TOYO0045359 TOYO0045360 TOYO0045361 TOYO0045362 TOYO0045363 TOYO0045364 TOYO0045365 TOYO0045366 TOYO0045367 TOYO0045368 TOYO0045369 TOYO0045370 TOYO0045371 TOYO0045372 TOYO0045373 TOYO0045374 TOYO0045375 TOYO0045376 TOYO0045377 TOYO0045378 TOYO0045379 TOYO0045380 TOYO0045381 TOYO0045382 TOYO0045383 TOYO0045384 TOYO0045385 TOYO0045386 TOYO0045387 TOYO0045388 TOYO0045389 TOYO0045390 TOYO0045392 TOYO0045393 TOYO0045394 TOYO0045395 TOYO0045396 TOYO0045397
SkillPak™ BIO Pre-Packed Columns
Columnas de cromatografía
SkillPak™ BIO pre-packed columns are designed for a fast method development and scale-up of multi-column chromatography (MCC) or continuous chromatography processes. They are pre-packed with TOYOPEARL® process chromatography media for bioseparations.
  • MCC-dedicated dimensions and packing procedures
  • Comprehensive resins for MCC applications
  • Multi-column kits with single resin lot and low column-to-column variability
They are ready to use and show excellent physical strength and ideal pressure-flow characteristic for downstrem processing in multi-column (More Product Information