Glassware Washer for Laboratories, TIVA8
Proveedor: TUTTNAUER
ATS238-0493
TUTTATS238-0493EA 0 EUR
TUTTATS238-0493
Glassware Washer for Laboratories, TIVA8
Lavadoras de vidrio
Tuttnauer's TIVA8 Lab line Washers contain unrivalled levels of efficiency that deliver on water and energy
savings. The high-quality system is specifically designed for research intensive laboratory settings that offer
automated cleaning and drying of typical laboratory glassware, as well as miscellaneous plastic items used in
research facilities.
  • Simple operation and process monitoring
  • Advanced control and warning system
  • Easy to clean and maintain process monitoring
  • Optimal results
  • Highest design standards
A wide range of racks, nozzles and accessoriesMore Product Information