Crucibles, porcelain
Proveedor: USBECK Laborgeräte
5665 5666 5667 5668
USBE5665EA 6 EUR
USBE5665 USBE5666 USBE5667 USBE5668
Crucibles, porcelain
Crisoles
These crucibles are made from glazed porcelain.
  • Temperature resistant up to 1000 °C
Certificaciones: DIN 12904