Anti-CD34 Mouse Monoclonal Antibody [clone: 132CT29.1.5]

Proveedor: US Biological
030848
USBI030848EA 667 EUR
USBI030848
Anti-CD34 Mouse Monoclonal Antibody [clone: 132CT29.1.5]
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: Monoclonal
Clone: 132CT29.1.5
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR