Anti-MB Mouse Monoclonal Antibody [clone: 444CT19.2.1]

Proveedor: US Biological
031150
USBI031150EA 667 EUR
USBI031150
Anti-MB Mouse Monoclonal Antibody [clone: 444CT19.2.1]
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:250-1000

Type: Primary
Antigen: MB (myoglobin)
Clonality: Monoclonal
Clone: 444CT19.2.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR