Anti-TNK1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 170CT7.5.4]

Proveedor: US Biological
031302-PE
USBI031302-PEEA 884 EUR
USBI031302-PE
Anti-TNK1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 170CT7.5.4]
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1000
Do not freeze! Light sensitive

Type: Primary
Antigen: TNK1 (tyrosine kinase, non-receptor, 1)
Clonality: Monoclonal
Clone: 170CT7.5.4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR