Anti-AMELX Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Proveedor: US Biological
031833-APC
USBI031833-APCEA 884 EUR
USBI031833-APC
Anti-AMELX Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: AMELX (amelogenin, X-linked)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR