Anti-ATF5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Proveedor: US Biological
032163-AP
USBI032163-APEA 884 EUR
USBI032163-AP
Anti-ATF5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ATF5 (activating transcription factor 5)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno ATF5
Environmentally Preferable
Nombre del antígeno Activating transcription factor 5
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado AP (Alkaline Phosphatase)
Reactividad Human
Huésped Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
Epítopo C-terminal
Número de aminoácidos 187 to 216
Forma liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR