Anti-BOLL Rabbit Polyclonal Antibody

Proveedor: US Biological
032559
USBI032559EA 667 EUR
USBI032559
Anti-BOLL Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: BOLL (boule-like RNA-binding protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR