Anti-ZGRF1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Proveedor: US Biological
032979-PE
USBI032979-PEEA 884 EUR
USBI032979-PE
Anti-ZGRF1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50
Do not freeze!

Type: Primary
Antigen: ZGRF1 (zinc finger, GRF-type containing 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

ESPECIFICACIONES

Simbolo antígeno ZGRF1
Environmentally Preferable
Nombre del antígeno zinc finger, GRF-type containing 1
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado PE (Phycoerythrin)
Reactividad Human,Mouse
Huésped Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
FluorescenciaInmunológica Yes
Epítopo N-terminal
Número de aminoácidos 45 to 74
Forma liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR