Anti-DHCR7 Rabbit Polyclonal Antibody

Proveedor: US Biological
034607
USBI034607EA 667 EUR
USBI034607
Anti-DHCR7 Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: DHCR7 (7-dehydrocholesterol reductase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR