Anti-GLB1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: US Biological
036066-FITC
USBI036066-FITCEA 884 EUR
USBI036066-FITC
Anti-GLB1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: GLB1 (galactosidase, beta 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR