Anti-His6-DEP-1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Proveedor: US Biological
036578-AP
USBI036578-APEA 884 EUR
USBI036578-AP
Anti-His6-DEP-1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50
Do not freeze!

Type: Primary
Antigen: His6-DEP-1
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno HIS6-DEP-1
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado AP (Alkaline Phosphatase)
Reactividad Human
Huésped Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
FluorescenciaInmunológica Yes
Forma liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR