Anti-MARCO Rabbit Polyclonal Antibody (Biotin)

Proveedor: US Biological
038200-BIOTIN
USBI038200-BIOTINEA 615 EUR
USBI038200-BIOTIN
Anti-MARCO Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: MARCO (macrophage receptor with collagenous structure)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR