Anti-OR4K1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Proveedor: US Biological
039398-APC
USBI039398-APCEA 884 EUR
USBI039398-APC
Anti-OR4K1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: OR4K1 (olfactory receptor, family 4, subfamily K, member 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR