Anti-PPP2R1A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: US Biological
040354-FITC
USBI040354-FITCEA 884 EUR
USBI040354-FITC
Anti-PPP2R1A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: PPP2R1A (protein phosphatase 2, regulatory subunit A, alpha)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno PPP2R1A
Nombre del antígeno protein phosphatase 2, regulatory subunit A, alpha
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado FITC (Fluorescein Isothiocyanate)
Reactividad Human
Huésped Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
Epítopo C-terminal
Número de aminoácidos 454 to 481
Forma liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Temperatura de almacenamiento Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR