Anti-RFT1 Rabbit Polyclonal Antibody (Biotin)

Proveedor: US Biological
040982-BIOTIN
USBI040982-BIOTINEA 884 EUR
USBI040982-BIOTIN
Anti-RFT1 Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: RFT1 (RFT1 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR