Anti-SEL1L Rabbit Polyclonal Antibody

Proveedor: US Biological
041527
USBI041527EA 667 EUR
USBI041527
Anti-SEL1L Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SEL1L (sel-1 suppressor of lin-12-like (C. elegans))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR