Anti-TAF3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: US Biological
042580-HRP
USBI042580-HRPEA 884 EUR
USBI042580-HRP
Anti-TAF3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: TAF3 (TAF3 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 140kDa)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR