Anti-ATRAID Mouse Monoclonal Antibody [clone: 1D2]

Proveedor: US Biological
123753
USBI123753EA 655 EUR
USBI123753
Anti-ATRAID Mouse Monoclonal Antibody [clone: 1D2]
Anticuerpos
Type: Primary
Antigen: ATRAID (all-trans retinoic acid-induced differentiation factor)
Clonality: monoclonal
Clone: 1D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno ATRAID
Environmentally Preferable
Nombre del antígeno all-trans retinoic acid-induced differentiation factor
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 1D2
Reactividad Human
Huésped Mouse
Isotipo IgG2a kappa
ELISA Yes
Sequence MLHARCCLNQKGTILGLDLQNCSLEDPGPNFHQAHTTVIIDLQANPLKGDLANTFRGFTQLQTLILPQHVNCPGGINAWNTITSYIDNQICQGQKNLCNNTGDPEMCPENGSCVPDGPGLLQCVCADGFHGYKCMRQGSFSLLMFFGILGATTLSVSILLWATQRRKAKTS
Forma liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2.
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR