Anti-CABP4 Mouse Monoclonal Antibody [clone: 5G11]

Proveedor: US Biological
124202
USBI124202EA 655 EUR
USBI124202
Anti-CABP4 Mouse Monoclonal Antibody [clone: 5G11]
Anticuerpos
Type: Primary
Antigen: CABP4 (calcium binding protein 4)
Clonality: monoclonal
Clone: 5G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno CABP4
Environmentally Preferable
Nombre del antígeno calcium binding Protein 4
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 5G11
Reactividad Human
Huésped Mouse
Isotipo IgG
Western blot Yes
ELISA Yes
Sequence MTEPWLALGTSWTLPLQDRELGPEELDELQAAFEEFDTDRDGYISHRELGDCMRTLGYMPTEMELLEVSQHIKMRMGGRVDFEEFVELIGPKLREETAHMLGVRELRIAFREFDRDRDGRITVAELREAVPALLGEPLAGPELDEMLREVDLNGDGTVDFDEFVMMLSRH
Forma liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2.
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR