Anti-CD7 Mouse Monoclonal Antibody [clone: 1B8]

Proveedor: US Biological
124675
USBI124675EA 655 EUR
USBI124675
Anti-CD7 Mouse Monoclonal Antibody [clone: 1B8]
Anticuerpos
Type: Primary
Antigen: CD7 (CD7 molecule)
Clonality: monoclonal
Clone: 1B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR