Anti-Choline Acetyltransferase Mouse Monoclonal Antibody [clone: 1H7]

Proveedor: US Biological
124965
USBI124965EA 655 EUR
USBI124965
Anti-Choline Acetyltransferase Mouse Monoclonal Antibody [clone: 1H7]
Anticuerpos
Type: Primary
Antigen: Choline Acetyltransferase
Clonality: monoclonal
Clone: 1H7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR