Anti-EFCAB6 Mouse Polyclonal Antibody

Proveedor: US Biological
126867
USBI126867EA 655 EUR
USBI126867
Anti-EFCAB6 Mouse Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: EFCAB6 (EF-hand calcium binding domain 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR