Anti-HAL Rabbit Polyclonal Antibody

Proveedor: US Biological
127714
USBI127714EA 696 EUR
USBI127714
Anti-HAL Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: HAL (histidine ammonia-lyase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR