Anti-LAMTOR3 Mouse Monoclonal Antibody [clone: 2A4]

Proveedor: US Biological
129420
USBI129420EA 623 EUR
USBI129420
Anti-LAMTOR3 Mouse Monoclonal Antibody [clone: 2A4]
Anticuerpos
Type: Primary
Antigen: LAMTOR3 (late endosomal/lysosomal adaptor, MAPK and MTOR activator 3)
Clonality: monoclonal
Clone: 2A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR